Learn More
3-Cyclopentylpropionic acid, 98%
Product Code. 11463717
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. 11463717
Brand: Thermo Scientific Alfa Aesar A18192.14
Request Bulk or Custom Format
CAS: 140-77-2 | C8H14O2 | 142.20 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 140-77-2
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00001392
InChI Key ZRPLANDPDWYOMZ-UHFFFAOYSA-N
Synonym 3-cyclopentylpropionic acid, cyclopentanepropanoic acid, cyclopentanepropionic acid, 3-cylcopentylpropanoic acid, cyclopentylpropionic acid, propionic acid, 3-cyclopentyl, 3-cyclopentyl-propionic acid, cypionic acid, 3-cyclopentylpropionicacid, cipionate
PubChem CID 8818
ChEBI CHEBI:50899
IUPAC Name 3-cyclopentylpropanoic acid
SMILES OC(=O)CCC1CCCC1
Specifications
Density 0.996
Boiling Point 130°C to 133°C (12 mmHg)
Flash Point 47°C (117°F)
Refractive Index 1.457
Quantity 25 g
UN Number UN1993
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material 3-Cyclopentylpropionic acid