Learn More
3,5-Dimethoxyphenol, 98%
Product Code. p-7026332
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. 11495946
Brand: Thermo Scientific Alfa Aesar B22283.09
Request Bulk or Custom Format
CAS: 500-99-2 | C8H10O3 | 154.17 g/mol
3,5-dimethoxyphenol is a marker for poisoning from Taxus baccata.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-dimethoxyphenol is a marker for poisoning from Taxus baccata.
Solubility
Soluble in methanol, ethanol, benzene, water (slightly), and chloroform.
Notes
Air and light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, light.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 500-99-2
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00008388
InChI Key XQDNFAMOIPNVES-UHFFFAOYSA-N
Synonym taxicatigenin, phenol, 3,5-dimethoxy, phloroglucinol dimethyl ether, 1-hydroxy-3,5-dimethoxybenzene, 3,5-dimethoxy phenol, unii-23uxw8136a, phenol,5-dimethoxy, 3,5-dimethoxy-phenol, pubchem16421, acmc-209kk2
PubChem CID 10383
IUPAC Name 3,5-dimethoxyphenol
SMILES COC1=CC(OC)=CC(O)=C1
Specifications
Melting Point 45°C to 47°C
Boiling Point 172°C to 175°C (17 mmHg)
Flash Point 78°C (173°F)
Quantity 10 g
Beilstein 1239156
Sensitivity Air and light sensitive
Solubility Information Soluble in methanol,ethanol,benzene,water (slightly),and chloroform.
Formula Weight 154.16
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxyphenol