accessibility menu, dialog, popup

UserName

CH3OOHOCH3

CAS RN 500-99-2

IUPAC Name: 3,5-dimethoxyphenol
Synonyms: 3,5-dimethoxyphenol
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: XQDNFAMOIPNVES-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC(O)=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)