Learn More
3,4-Dihydroxybenzhydrazide, 97%
Product Code. 11490357
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490357 5 g 5g
11400367 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11490357 Supplier Thermo Scientific Alfa Aesar Supplier No. B21746.06
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
CAS: 39635-11-5 | C7H8N2O3 | 168.152 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 39635-11-5
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.152
MDL Number MFCD00017064
InChI Key WGXWEXNJRZMIPT-UHFFFAOYSA-N
Synonym 3,4-dihydroxybenzhydrazide, 3,4-dihydroxy-benzoic acid hydrazide, 4-???benzene-1,2-diol, protocatechuhydrazide, acmc-1ad2t, 3,4-dihydroxy-benzohydrazide, 3,4-bis oxidanyl benzohydrazide, 3,4-dihydroxybenzoic acid hydrazide, 3,4-dihydroxy-benzoesaeure-hydrazid, 3,4-dihydroxybenzoic acid, hydrazide
PubChem CID 586300
IUPAC Name 3,4-dihydroxybenzohydrazide
SMILES C1=CC(=C(C=C1C(=O)NN)O)O
Specifications
Melting Point ∼265°C (decomposition)
Quantity 5 g
Beilstein 2642968
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material 3,4-Dihydroxybenzhydrazide