accessibility menu, dialog, popup

UserName

NH2HNOOHHO

CAS RN 39635-11-5

IUPAC Name: 3,4-dihydroxybenzohydrazide
Synonyms: 3,4-dihydroxybenzohydrazide
Molecular Weight (g/mol): 168.15
Molecular Formula: C7H8N2O3
InChi Key: WGXWEXNJRZMIPT-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(O)C(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results