missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methoxyphenyl)propionic acid, 98+%

Product Code. 11426857
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11426857

Brand: Thermo Scientific Alfa Aesar A14314.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6342-77-4 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6342-77-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00002772
InChI Key XSZSNLOPIWWFHS-UHFFFAOYSA-N
Synonym 3-2-methoxyphenyl propanoic acid, 3-2-methoxyphenyl propionic acid, 2-methoxyhydrocinnamic acid, benzenepropanoic acid, 2-methoxy, o-methoxyhydrocinnamic acid, 3-o-methoxyphenyl propionic acid, 2-methoxy-benzenepropanoic acid, beta-o-methoxyphenyl propionic acid, acmc-209nfi, maybridge1_000611
PubChem CID 80652
IUPAC Name 3-(2-methoxyphenyl)propanoic acid
SMILES COC1=CC=CC=C1CCC(=O)O

Specifications

Melting Point 85°C to 88°C
Quantity 25 g
Beilstein 2097012
Formula Weight 180.2
Percent Purity ≥98%
Chemical Name or Material 3-(2-Methoxyphenyl)propionic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.