Learn More
2-Phenylethyl acetate, 98%
Product Code. 11434757
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
Product Code. 11434757
Brand: Thermo Scientific Alfa Aesar B21238.0B
Request Bulk or Custom Format
CAS: 103-45-7 | C10H12O2 | 164.20 g/mol
Phenylethyl acetate mantains the typical rose notes of Phenylethanol with a more intense honey profile which is perfect for fruit and alcoholic drink applications. It is used as a pharmaceutical intermediate and also it holds application in gas chromatography and liquid chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenylethyl acetate mantains the typical rose notes of Phenylethanol with a more intense honey profile which is perfect for fruit and alcoholic drink applications. It is used as a pharmaceutical intermediate and also it holds application in gas chromatography and liquid chromatography.
Solubility
It is insoluble in paraffin oil, water. Soluble in alcohol, dipropylene glycol, fixed oils, kerosene, propylene glycol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Protected from heat and light.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 103-45-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008720
InChI Key MDHYEMXUFSJLGV-UHFFFAOYSA-N
Synonym phenethyl acetate, 2-phenethyl acetate, acetic acid, 2-phenylethyl ester, benzylcarbinyl acetate, beta-phenylethyl acetate, acetic acid, phenethyl ester, phenethyl alcohol, acetate, phenylethyl acetate, acetic acid phenethyl ester, ethanol, 2-phenyl-, acetate
PubChem CID 7654
ChEBI CHEBI:31988
IUPAC Name 2-phenylethyl acetate
SMILES CC(=O)OCCC1=CC=CC=C1
Specifications
Melting Point -31°C
Density 1.029
Boiling Point 231°C to 233°C
Flash Point 107°C (225°F)
Odor Fruit-like
Refractive Index 1.499
Quantity 1000 g
Beilstein 638179
Solubility Information It is insoluble in paraffin oil,water. Soluble in alcohol,dipropylene glycol,fixed oils,kerosene,propylene glycol.
Formula Weight 164.21
Percent Purity 98%
Chemical Name or Material 2-Phenylethyl acetate