missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,5-dinitrobenzotrifluoride, 98+%

Product Code. p-7023751
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11438247

Brand: Thermo Scientific Alfa Aesar A17617.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 392-95-0 | C7H2ClF3N2O4 | 270.55 g/mol

It is used to increase mutagenic activity in the glutathione transferase mutant (C36).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to increase mutagenic activity in the glutathione transferase mutant (C36).

Solubility
Insoluble in water. Soluble in Chloroform, Methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry place. Store under 20˚C Freezer, Under Inert Atmosphere.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392-95-0
Molecular Formula C7H2ClF3N2O4
Molecular Weight (g/mol) 270.55
MDL Number MFCD00007076
InChI Key RLXKADBMLQPLDV-UHFFFAOYSA-N
Synonym 2-chloro-3,5-dinitrobenzotrifluoride, 2-chloro-1,5-dinitro-3-trifluoromethyl benzene, benzene, 2-chloro-1,5-dinitro-3-trifluoromethyl, 2-chloro-3,5-binitrotrifluorotoluene, pubchem4846, 2-chlor-1,5-dinitro-3-trifluormethyl benzol, ksc914e2d, labotest-bb lt00159813, labotest-bb lt03380099, 2-chloro-3,5-dinitro-benzotrifluoride
PubChem CID 67851
IUPAC Name 2-chloro-1,5-dinitro-3-(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC(=C(Cl)C(=C1)C(F)(F)F)[N+]([O-])=O

Specifications

Melting Point 61°C to 64°C
Quantity 5 g
UN Number UN2923
Beilstein 2061156
Solubility Information Insoluble in water. Soluble in Chloroform,Methanol.
Formula Weight 270.55
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-3,5-dinitrobenzotrifluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.