accessibility menu, dialog, popup

UserName

CAS RN 392-95-0

ONOClFFFNOO

CAS RN 392-95-0

IUPAC Name: 2-chloro-1,5-dinitro-3-(trifluoromethyl)benzene
Synonyms: 2-chloro-1,5-dinitro-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 270.55
Molecular Formula: C7H2ClF3N2O4
InChi Key: RLXKADBMLQPLDV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=C(Cl)C(=C1)C(F)(F)F)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results