missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromopropionamide, 97%, Thermo Scientific™

Product Code. 11313116
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313116 25 g 25g
11303116 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313116 Supplier Thermo Scientific Alfa Aesar Supplier No. L10978.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,3-Dibromopropionamide, is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15102-42-8
Molecular Formula C3H5Br2NO
Molecular Weight (g/mol) 230.887
MDL Number MFCD00031484
InChI Key DZQCMQRQFZXQKN-UHFFFAOYSA-N
PubChem CID 85798
IUPAC Name 2,3-dibromopropanamide
SMILES C(C(C(=O)N)Br)Br

Specifications

Melting Point 132°C to 133°C
Quantity 25 g
Beilstein 1749448
Synonym 2,3-dibromopropionamide, propanamide, 2,3-dibromo, acmc-209twc, propionamide,3-dibromo, 2,3-dibromo-propionamide, #, propanamide,2,3-dibromo, propionamide, 2,3-dibromo, ksc493i7n, 2,3-dibromopropionamide standardforacrylamideglcdetermination
Solubility Information Insoluble in water.
Formula Weight 230.9
Percent Purity 97%
Chemical Name or Material 2,3-Dibromopropionamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.