accessibility menu, dialog, popup

UserName

CAS RN 15102-42-8

NH2OBrBr

CAS RN 15102-42-8

IUPAC Name: 2,3-dibromopropanamide
Synonyms: 2,3-dibromopropanamide
Molecular Weight (g/mol): 230.89
Molecular Formula: C3H5Br2NO
InChi Key: DZQCMQRQFZXQKN-UHFFFAOYNA-N
SMILES: NC(=O)C(Br)CBr
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results