Learn More
2,3-Dibromopropionamide, 97%, Thermo Scientific™
Product Code. 11303116
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303116 5 g 5g
11313116 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11303116 Supplier Thermo Scientific Alfa Aesar Supplier No. L10978.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2,3-Dibromopropionamide, is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15102-42-8
Molecular Formula C3H5Br2NO
Molecular Weight (g/mol) 230.887
MDL Number MFCD00031484
InChI Key DZQCMQRQFZXQKN-UHFFFAOYSA-N
PubChem CID 85798
IUPAC Name 2,3-dibromopropanamide
SMILES C(C(C(=O)N)Br)Br
Specifications
Melting Point 132°C to 133°C
Quantity 5 g
Beilstein 1749448
Synonym 2,3-dibromopropionamide, propanamide, 2,3-dibromo, acmc-209twc, propionamide,3-dibromo, 2,3-dibromo-propionamide, #, propanamide,2,3-dibromo, propionamide, 2,3-dibromo, ksc493i7n, 2,3-dibromopropionamide standardforacrylamideglcdetermination
Solubility Information Insoluble in water.
Formula Weight 230.9
Percent Purity 97%
Chemical Name or Material 2,3-Dibromopropionamide