missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Aminocarbonyl)-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid, TRC

Product Code. 30135961
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135961 100 mg 100mg
30009868 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135961 Supplier Toronto Research Chemicals Supplier No. TRCA603115100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1239233-86-3
Molecular Formula C16 H18 N2 O4 S
Molecular Weight (g/mol) 334.39
Synonym 2-[3-(Aminocarbonyl)-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid, 2-(3-Carbamoyl-4-isobutoxyphenyl)-4-methylthiazole-5-carboxylic Acid
IUPAC Name 2-[3-carbamoyl-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC(C)COc1ccc(cc1C(=O)N)c2nc(C)c(s2)C(=O)O

Specifications

Analyte or Component Names 2-[3-(Aminocarbonyl)-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid
Shipping Condition Room Temperature
InChI Formula InChI=1S/C16H18N2O4S/c1-8(2)7-22-12-5-4-10(6-11(12)14(17)19)15-18-9(3)13(23-15)16(20)21/h4-6,8H,7H2,1-3H3,(H2,17,19)(H,20,21)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 mg
Formula Weight 334.0987
Percent Purity >95%
Chemical Name or Material 2-[3-(Aminocarbonyl)-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.