CAS RN 1239233-86-3
CAS RN 1239233-86-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-[3-(Aminocarbonyl)-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid, TRC
CAS: 1239233-86-3 Molecular Formula: C16 H18 N2 O4 S Molecular Weight (g/mol): 334.39 Synonym: 2-[3-(Aminocarbonyl)-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid,2-(3-Carbamoyl-4-isobutoxyphenyl)-4-methylthiazole-5-carboxylic Acid IUPAC Name: 2-[3-carbamoyl-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylic acid SMILES: CC(C)COc1ccc(cc1C(=O)N)c2nc(C)c(s2)C(=O)O