missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ 11 beta-HSD1 Antibody (SR3601), Novus Biologicals™
Rabbit Monoclonal Antibody
339.00€ - 406.00€
Specifications
| Antigen | 11 beta-HSD1 |
|---|---|
| Clone | SR3601 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041589
|
Novus Biologicals™
NBP3-27783-50ul |
50 μL |
339.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041787
|
Novus Biologicals™
NBP3-27783-100ul |
100 μL |
406.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
11 beta-HSD1 Monoclonal antibody specifically detects 11 beta-HSD1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| 11 beta-HSD1 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 11-beta-hydroxysteroid dehydrogenase 1, EC 1.1.1, EC 1.1.1.146, HDL, HSD11HSD11B, HSD11LMGC13539, hydroxysteroid (11-beta) dehydrogenase 1, member 1, SDR26C1 | |
| A synthesized peptide derived from human 11 beta-HSD1 (Uniprot #: P28845) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3601 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 3290 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title