missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ 11 beta-HSD1 Antibody (SR3601), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27783-100ul
This item is not returnable.
View return policy
Description
11 beta-HSD1 Monoclonal antibody specifically detects 11 beta-HSD1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| 11 beta-HSD1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot | |
| SR3601 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| 11-beta-hydroxysteroid dehydrogenase 1, EC 1.1.1, EC 1.1.1.146, HDL, HSD11HSD11B, HSD11LMGC13539, hydroxysteroid (11-beta) dehydrogenase 1, member 1, SDR26C1 | |
| A synthesized peptide derived from human 11 beta-HSD1 (Uniprot #: P28845) | |
| 100 μL | |
| Lipid and Metabolism | |
| 3290 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction