missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A20/TNFAIP3 Mouse anti-Human, Clone: TNFAIP3/2813, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07730-100ug
This item is not returnable.
View return policy
Description
A20/TNFAIP3 Monoclonal specifically detects A20/TNFAIP3 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| A20/TNFAIP3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| A20TNFA1P2, EC 3.4.19.12, EC 6.3.2.-, MGC104522, MGC138687, OTU domain-containing protein 7C, OTUD7CMGC138688, Putative DNA-binding protein A20, TNF alpha-induced protein 3, tumor necrosis factor alpha-induced protein 3, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha-induced protein 3, Zinc finger protein A20 | |
| Recombinant fragment (around aa 241-356) of human A20/TNFAIP3 protein (exact sequence is proprietary) | |
| 100 μg | |
| Cancer, Inflammation, Innate Immunity, Necroptosis, Neurodegeneration, Signal Transduction | |
| 7128 | |
| Store at 4°C. | |
| IgG2c κ |
| Immunohistochemistry (Paraffin) | |
| TNFAIP3/2813 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction