missing translation for 'onlineSavingsMsg'
Learn More

A20/TNFAIP3 Mouse anti-Human, Clone: TNFAIP3/2813, Novus Biologicals™

Mouse Monoclonal Antibody

484.00€

Specifications

Antigen A20/TNFAIP3
Clone TNFAIP3/2813
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 1 to 2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18676596
View Documents
Novus Biologicals
NBP3-07730-100ug
100 μg
484.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A20/TNFAIP3 Monoclonal specifically detects A20/TNFAIP3 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

A20/TNFAIP3
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer, Inflammation, Innate Immunity, Necroptosis, Neurodegeneration, Signal Transduction
10 mM PBS with 0.05% BSA
7128
IgG2c κ
Protein A or G purified
TNFAIP3/2813
Immunohistochemistry-Paraffin 1 to 2 μg/mL
Monoclonal
Purified
RUO
Human
A20TNFA1P2, EC 3.4.19.12, EC 6.3.2.-, MGC104522, MGC138687, OTU domain-containing protein 7C, OTUD7CMGC138688, Putative DNA-binding protein A20, TNF alpha-induced protein 3, tumor necrosis factor alpha-induced protein 3, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha-induced protein 3, Zinc finger protein A20
Recombinant fragment (around aa 241-356) of human A20/TNFAIP3 protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.