Learn More
NanoLuc™ (Nluc) Luciferase Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB100261-100
 View more versions of this product
Product Code. 18793607
  • ONLINE EXCLUSIVE PRICE
    490.00€
    463.05€ / 100µg
    Save 26.95€
    5% Off
Description
Luciferase Monoclonal specifically detects Luciferase in Multi-Species samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Luciferase
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG2a
Western Blot
965853
Western Blot 2 μg/mL
ec 1.13.12.7, Fluc, luciferin 4 monooxygenase
Synthetic peptide for NanoLuc™ (Nluc) Luciferase
100 μg
Detects NanoLuc™ (Nluc) Luciferase in direct ELISAs. In Western blots, no cross-reactivity with Large BiT is observed.
Multi-species
Supernatant