CAS RN 96382-71-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Dehydro Felodipine, TRC
CAS: 96382-71-7 Molecular Formula: C18 H17 Cl2 N O4 Molecular Weight (g/mol): 382.24 Synonym: Ethyl Methyl 4-(2,3-Dichlorophenyl)-2,6-dimethylpyridine-3,5-dicarboxylate,Felodipine Imp. A (EP),3,5-Pyridinedicarboxylic acid, 4-(2,3-dichlorophenyl)-2,6-dimethyl-, ethyl methyl ester (9CI),Dehydrofelodipine,H 152/37 IUPAC Name: 5-O-ethyl 3-O-methyl 4-(2,3-dichlorophenyl)-2,6-dimethylpyridine-3,5-dicarboxylate SMILES: CCOC(=O)c1c(C)nc(C)c(C(=O)OC)c1c2cccc(Cl)c2Cl
2
Ethyl Methyl 4-(2,3-Dichlorophenyl)-2,6-dimethylpyridine-3,5-dicarboxylate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.