Learn More
Ethyl Methyl 4-(2,3-Dichlorophenyl)-2,6-dimethylpyridine-3,5-dicarboxylate, Mikromol™
Product Code. 17971626
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971626 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17971626 Supplier Mikromol Supplier No. MM0139.01
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl Methyl 4-(2,3-Dichlorophenyl)-2,6-dimethylpyridine-3,5-dicarboxylate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethyl Methyl 4-(2,3-Dichlorophenyl)-2,6-dimethylpyridine-3,5-dicarboxylate
CAS 96382-71-7
Analyte or Component Names Ethyl Methyl 4-(2,3-Dichlorophenyl)-2,6-dimethylpyridine-3,5-dicarboxylate
Shipping Condition Room Temperature
Molecular Formula C18 H17 Cl2 N O4
Certifications/Compliance ISO 9001
Synonym Felodipine Imp. A (EP), Felodipine USP Related Compound A, Felodipine USP RC A, Ethyl Methyl 4-(2,3-Dichlorophenyl)-2,6-dimethylpyridine-3,5-dicarboxylate, 3,5-Pyridinedicarboxylic acid, 4-(2,3-dichlorophenyl)-2,6-dimethyl-, ethyl methyl ester (9CI), Dehydrofelodipine, H 152/37, Felodipine Related Compound A, Felodipine Impurity A
InChI Formula InChI=1S/C18H17Cl2NO4/c1-5-25-18(23)14-10(3)21-9(2)13(17(22)24-4)15(14)11-7-6-8-12(19)16(11)20/h6-8H,5H2,1-4H3
SMILES CCOC(=O)c1c(C)nc(C)c(C(=O)OC)c1c2cccc(Cl)c2Cl
IUPAC Name 5-O-ethyl 3-O-methyl 4-(2,3-dichlorophenyl)-2,6-dimethylpyridine-3,5-dicarboxylate
Molecular Weight (g/mol) 382.24
Quantity 50 mg
Formula Weight 381.0535 g/mol