Learn More
Dehydro Felodipine, TRC
Product Code. 30135229
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135229 5 mg 5mg
30133715 1 mg 1mg
30009061 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135229 Supplier Toronto Research Chemicals Supplier No. TRCD2296505MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 96382-71-7
Molecular Formula C18 H17 Cl2 N O4
Molecular Weight (g/mol) 382.24
Synonym Ethyl Methyl 4-(2,3-Dichlorophenyl)-2,6-dimethylpyridine-3,5-dicarboxylate, Felodipine Imp. A (EP), 3,5-Pyridinedicarboxylic acid, 4-(2,3-dichlorophenyl)-2,6-dimethyl-, ethyl methyl ester (9CI), Dehydrofelodipine, H 152/37
IUPAC Name 5-O-ethyl 3-O-methyl 4-(2,3-dichlorophenyl)-2,6-dimethylpyridine-3,5-dicarboxylate
SMILES CCOC(=O)c1c(C)nc(C)c(C(=O)OC)c1c2cccc(Cl)c2Cl
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C18H17Cl2NO4/c1-5-25-18(23)14-10(3)21-9(2)13(17(22)24-4)15(14)11-7-6-8-12(19)16(11)20/h6-8H,5H2,1-4H3
Formula Weight 381.0535
Percent Purity >95%
Chemical Name or Material Ethyl Methyl 4-(2,3-Dichlorophenyl)-2,6-dimethylpyridine-3,5-dicarboxylate
Grade HPLC