CAS RN 81131-70-6
CAS RN 81131-70-6
IUPAC Name:
sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
Synonyms:
pravastatin sodium
Molecular Weight (g/mol):
446.52
Molecular Formula:
C23H35NaO7
InChi Key:
VWBQYTRBTXKKOG-IYNICTALSA-M
SMILES:
[Na+].CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC([O-])=O)[C@@H]12
1
–
3
of
3
results
2
Pravastatin Sodium, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Pravastatin Sodium, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.