missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pravastatin Sodium, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Pravastatin Sodium |
| CAS | 81131-70-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C23 H35 O7 . Na |
| Quantity | 50 mg |
| Synonym | Pravastatin sodium, (βR,ΔR,1S,2S,6S,8S,8aR)-1,2,6,7,8,8a-Hexahydro-β,Δ,6-trihydroxy-2-methyl-8-[(2S)-2-methyl-1-oxobutoxy]-1-naphthaleneheptanoic acid monosodium salt, sodium (+)-(3R,5R)-3,5-dihydroxy-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-[(S)-2-methylbutyryloxy]-1,2,6,7,8,8a-hexahydro-1-naphthyl]heptanoate, Eptastatin sodium, 3β-Hydroxycompactin sodium salt, 1-Naphthaleneheptanoic acid','1,2,6,7,8,8a-hexahydro-β,Δ,6-trihydroxy-2-methyl-8-[(2S)-2-methyl-1-oxobutoxy]-','sodium salt (1:1)','(βR,ΔR,1S,2S,6S,8S,8aR)-, 1-Naphthaleneheptanoic acid','1,2,6,7,8,8a-hexahydro-β,Δ,6-trihydroxy-2-methyl-8-(2-methyl-1-oxobutoxy)-','monosodium salt','[1S-[1α(βS*,ΔS*),2α,6α,8β(R*),8aα]]-, 1-Naphthaleneheptanoic acid','1,2,6,7,8,8a-hexahydro-β,Δ,6-trihydroxy-2-methyl-8-[(2S)-2-methyl-1-oxobutoxy]-','monosodium salt','(βR,ΔR,1S,2S,6S,8S,8aR)- (9CI), CS 514, Elisor, Epastatin sodium, Lipidal, Lipostat, Liprevil, Mevalotin, Mevan, Oliprevin, Prava, Pravachol, Pravacol, Pravaselect, Pravasin, Pravasine, Pravastatin sodium salt, Pravastin, Pravator, SQ 31000, Selectin, Selektine, Selipran, Sodium pravastatin, Vasten |
| InChI Formula | InChI=1S/C23H36O7.Na/c1-4-13(2)23(29)30-20-11-17(25)9-15-6-5-14(3)19(22(15)20)8-7-16(24)10-18(26)12-21(27)28;/h5-6,9,13-14,16-20,22,24-26H,4,7-8,10-12H2,1-3H3,(H,27,28);/q;+1/p-1/t13-,14-,16+,17+,18+,19-,20-,22-;/m0./s1 |
| SMILES | [Na+].CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)[O-])[C@@H]12 |
| IUPAC Name | sodium;(3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-[(2S)-2-methylbutanoyl]oxy-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate |
| Molecular Weight (g/mol) | 446.51 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction