missing translation for 'onlineSavingsMsg'
Learn More

Pravastatin Sodium, TRC

Product Code. 30409733
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30409733

Brand: Toronto Research Chemicals TRCP70200025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pravastatin Sodium
CAS 81131-70-6
Purity Grade Notes HPLC
Molecular Formula C23 H35 O7 . Na
Quantity 25 mg
Synonym Pravastatin sodium, (βR,ΔR,1S,2S,6S,8S,8aR)-1,2,6,7,8,8a-Hexahydro-β,Δ,6-trihydroxy-2-methyl-8-[(2S)-2-methyl-1-oxobutoxy]-1-naphthaleneheptanoic acid monosodium salt, sodium (+)-(3R,5R)-3,5-dihydroxy-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-[(S)-2-methylbutyryloxy]-1,2,6,7,8,8a-hexahydro-1-naphthyl]heptanoate, Eptastatin sodium, 3β-Hydroxycompactin sodium salt, 1-Naphthaleneheptanoic acid','1,2,6,7,8,8a-hexahydro-β,Δ,6-trihydroxy-2-methyl-8-[(2S)-2-methyl-1-oxobutoxy]-','sodium salt (1:1)','(βR,ΔR,1S,2S,6S,8S,8aR)-, 1-Naphthaleneheptanoic acid','1,2,6,7,8,8a-hexahydro-β,Δ,6-trihydroxy-2-methyl-8-(2-methyl-1-oxobutoxy)-','monosodium salt','[1S-[1α(βS*,ΔS*),2α,6α,8β(R*),8aα]]-, 1-Naphthaleneheptanoic acid','1,2,6,7,8,8a-hexahydro-β,Δ,6-trihydroxy-2-methyl-8-[(2S)-2-methyl-1-oxobutoxy]-','monosodium salt','(βR,ΔR,1S,2S,6S,8S,8aR)- (9CI), CS 514, Elisor, Epastatin sodium, Lipidal, Lipostat, Liprevil, Mevalotin, Mevan, Oliprevin, Prava, Pravachol, Pravacol, Pravaselect, Pravasin, Pravasine, Pravastatin sodium salt, Pravastin, Pravator, SQ 31000, Selectin, Selektine, Selipran, Sodium pravastatin, Vasten
InChI Formula InChI=1S/C23H36O7.Na/c1-4-13(2)23(29)30-20-11-17(25)9-15-6-5-14(3)19(22(15)20)8-7-16(24)10-18(26)12-21(27)28;/h5-6,9,13-14,16-20,22,24-26H,4,7-8,10-12H2,1-3H3,(H,27,28);/q;+1/p-1/t13-,14-,16+,17+,18+,19-,20-,22-;/m0./s1
SMILES [Na+].CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)[O-])[C@@H]12
IUPAC Name sodium;(3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-[(2S)-2-methylbutanoyl]oxy-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
Molecular Weight (g/mol) 446.51
Formula Weight 446.228
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.