accessibility menu, dialog, popup

UserName

CAS RN 81129-83-1

NaH3CH3C(S)ONHS(S)NH2OHOOO(Z)

CAS RN 81129-83-1

IUPAC Name: sodium (2Z)-7-{[(2S)-2-amino-2-carboxyethyl]sulfanyl}-2-{[(1S)-2,2-dimethylcyclopropyl]formamido}hept-2-enoate
Synonyms: cilastatin
Molecular Weight (g/mol): 380.43
Molecular Formula: C16H25N2NaO5S
InChi Key: QXPBTTUOVWMPJN-VQAYNFQASA-M
SMILES: [Na+].CC1(C)C[C@@H]1C(=O)N\C(=C/CCCCSC[C@@H](N)C(O)=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results