missing translation for 'onlineSavingsMsg'
Learn More

Cilastatin Sodium, Mikromol™

Product Code. 17951306
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17951306

Brand: Mikromol MM1092.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Cilastatin Sodium
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cilastatin sodium
CAS 81129-83-1
Analyte or Component Names Cilastatin sodium
Shipping Condition Room Temperature
Molecular Formula C16 H25 N2 O5 S . Na
Certifications/Compliance ISO 17034
Synonym Cilastatin sodium, Sodium (Z)-7-[[(R)-2-amino-2-carboxyethyl]sulfanyl]-2-[[[(1S)-2,2-dimethylcyclopropyl]carbonyl]amino]hept-2-enoate, 2-Heptenoic acid, 7-[[(2R)-2-amino-2-carboxyethyl]thio]-2-[[[(1S)-2,2-dimethylcyclopropyl]carbonyl]amino]-, sodium salt (1:1), (2Z)-, 2-Heptenoic acid, 7-[(2-amino-2-carboxyethyl)thio]-2-[[(2,2-dimethylcyclopropyl)carbonyl]amino]-, monosodium salt, [R-[R*,S*-(Z)]]-, 2-Heptenoic acid, 7-[[(2R)-2-amino-2-carboxyethyl]thio]-2-[[[(1S)-2,2-dimethylcyclopropyl]carbonyl]amino]-, monosodium salt, (2Z)- (9CI), Cilastatin sodium salt, L 642957, MK 791
InChI Formula InChI=1S/C16H26N2O5S.Na/c1-16(2)8-10(16)13(19)18-12(15(22)23)6-4-3-5-7-24-9-11(17)14(20)21;/h6,10-11H,3-5,7-9,17H2,1-2H3,(H,18,19)(H,20,21)(H,22,23);/q;+1/p-1/b12-6-;/t10-,11+;/m1./s1
SMILES [Na+].CC1(C)C[C@@H]1C(=O)N\C(=C/CCCCSC[C@H](N)C(=O)O)\C(=O)[O-]
IUPAC Name sodium;(Z)-7-[(2R)-2-amino-2-carboxyethyl]sulfanyl-2-[[(1S)-2,2-dimethylcyclopropanecarbonyl]amino]hept-2-enoate
Molecular Weight (g/mol) 380.43
Quantity 250 mg
Formula Weight 380.1382 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.