CAS RN 73003-80-2
CAS RN 73003-80-2
IUPAC Name:
dibromopropanediamide
Synonyms:
dibromopropanediamide
Molecular Weight (g/mol):
259.89
Molecular Formula:
C3H4Br2N2O2
InChi Key:
SWHQVMGRXIYDSF-UHFFFAOYSA-N
SMILES:
NC(=O)C(Br)(Br)C(N)=O
1
–
1
of
1
results
1
Dibromomalonamide, 98+%
CAS: 73003-80-2 Molecular Formula: C3H4Br2N2O2 Molecular Weight (g/mol): 259.89 MDL Number: MFCD00014803 InChI Key: SWHQVMGRXIYDSF-UHFFFAOYSA-N PubChem CID: 175375 SMILES: NC(=O)C(Br)(Br)C(N)=O