accessibility menu, dialog, popup

UserName

CAS RN 71989-20-3

H2NONHOOOHO

CAS RN 71989-20-3

IUPAC Name: 4-carbamoyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: 4-carbamoyl-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol): 368.39
Molecular Formula: C20H20N2O5
InChi Key: IZKGGDFLLNVXNZ-UHFFFAOYNA-N
SMILES: NC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results