Learn More
Nalpha-Fmoc-L-glutamine, 95%, Thermo Scientific™
Product Code. 15445388
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15445388
Brand: Thermo Scientific Alfa Aesar H59159.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 71989-20-3
Molecular Formula C20H20N2O5
Molecular Weight (g/mol) 368.39
MDL Number MFCD00037137
InChI Key IZKGGDFLLNVXNZ-UHFFFAOYNA-N
Synonym fmoc-gln-oh, nalpha-fmoc-l-glutamine, fmoc-l-gln-oh, fmoc-l-glutamine, n-fmoc-l-glutamine, fmoc-dap ac-oh, fmoc-gln, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-amino-5-oxopentanoic acid, 2s-4-carbamoyl-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n alpha-fluorenylmethoxycarbonylglutamine
PubChem CID 2724775
SMILES NC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Specifications
Melting Point 224°C (decomposition)
Quantity 5 g
Beilstein 4722773
Optical Rotation −19° (c=1 in DMF)
IUPAC Name 4-carbamoyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Formula Weight 368.38
Percent Purity 95%
Chemical Name or Material Nalpha-Fmoc-L-glutamine