CAS RN 66-84-2
CAS RN 66-84-2
IUPAC Name:
hydrogen 2-amino-3,4,5,6-tetrahydroxyhexanal chloride
Synonyms:
glucosamine hcl
Molecular Weight (g/mol):
215.63
Molecular Formula:
C6H14ClNO5
InChi Key:
CBOJBBMQJBVCMW-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].NC(C=O)C(O)C(O)C(O)CO
1
–
8
of
8
results
1
Promotions Available
Thermo Scientific Chemicals D-Glucosamine hydrochloride, 98+%
Thermo Scientific Chemicals D-Glucosamine hydrochloride, 98+%
CAS: 66-84-2 Molecular Formula: C6H13NO5·HCl
2
Thermo Scientific Chemicals D-Glucosamine hydrochloride, 98+%
CAS: 66-84-2 Molecular Formula: C6H13NO5·HCl MDL Number: MFCD00067674
7
D-Glucosamine Hydrochloride, TRC
CAS: 66-84-2 Molecular Formula: C6 H13 N O5 . Cl H Molecular Weight (g/mol): 215.63 Synonym: Glucosamine hydrochloride,2-Amino-2-deoxy-d-glucopyranose hydrochloride IUPAC Name: (3R,4R,5S,6R)-3-amino-6-(hydroxymethyl)oxane-2,4,5-triol;hydrochloride SMILES: Cl.N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O
8
Glucosamine Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.