Learn More
D-Glucosamine Hydrochloride, TRC
Product Code. 30005871
Change view
Click to view available options
Quantity:
10 mg
10 g
25 g
Unit Size:
10g
10mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005871 10 g 10g
17939434 10 mg 10mg
17947824 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30005871 Supplier Toronto Research Chemicals Supplier No. TRCG51495010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 66-84-2
Molecular Formula C6 H13 N O5 . Cl H
Molecular Weight (g/mol) 215.63
Synonym Glucosamine hydrochloride, 2-Amino-2-deoxy-d-glucopyranose hydrochloride
IUPAC Name (3R,4R,5S,6R)-3-amino-6-(hydroxymethyl)oxane-2,4,5-triol;hydrochloride
SMILES Cl.N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O
Specifications
Quantity 10 g
InChI Formula InChI=1S/C6H13NO5.ClH/c7-3-5(10)4(9)2(1-8)12-6(3)11;/h2-6,8-11H,1,7H2;1H/t2-,3-,4-,5-,6?;/m1./s1
Formula Weight 215.0561
Percent Purity >95%
Chemical Name or Material D-(+)-Glucosamine hydrochloride
Grade HPLC