Learn More
Glucosamine Hydrochloride, Mikromol™
Product Code. 17929014
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17929014 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17929014 Supplier Mikromol Supplier No. MM0695.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Glucosamine Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material D-(+)-Glucosamine hydrochloride
CAS 66-84-2
Analyte or Component Names D-(+)-Glucosamine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C6 H13 N O5 . Cl H
Certifications/Compliance ISO 17025
Synonym Glucosamine hydrochloride, 2-Amino-2-deoxy-d-glucopyranose hydrochloride
InChI Formula InChI=1S/C6H13NO5.ClH/c7-3-5(10)4(9)2(1-8)12-6(3)11;/h2-6,8-11H,1,7H2;1H/t2-,3-,4-,5-,6?;/m1./s1
SMILES Cl.N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O
IUPAC Name (3R,4R,5S,6R)-3-amino-6-(hydroxymethyl)oxane-2,4,5-triol;hydrochloride
Molecular Weight (g/mol) 215.63
Quantity 250 mg
Formula Weight 215.0561 g/mol