accessibility menu, dialog, popup

UserName

CAS RN 61903-11-5

H3CONHN

CAS RN 61903-11-5

IUPAC Name: 1-(1,4-diazepan-1-yl)ethan-1-one
Synonyms: 1-(1,4-diazepan-1-yl)ethanone
Molecular Weight (g/mol): 142.2
Molecular Formula: C7H14N2O
InChi Key: TWJPZMYNUBAUGA-UHFFFAOYSA-N
SMILES: CC(=O)N1CCCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results