accessibility menu, dialog, popup

UserName

ClNNNONHOHN

CAS RN 57808-66-9

IUPAC Name: 5-chloro-1-{1-[3-(2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl)propyl]piperidin-4-yl}-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: domperidone domperidon hexal gamir, domperidona Domperidon-TEVA apotex brand of domperidone maleate ratiopharm brand of domperidone maleate aliud brand of domperidone maleate domperidone monohydrochloride lopac-D-122 gastrocure
Molecular Weight (g/mol): 425.92
Molecular Formula: C22H24ClN5O2
InChi Key: FGXWKSZFVQUSTL-UHFFFAOYSA-N
SMILES: ClC1=CC=C2N(C3CCN(CCCN4C(=O)NC5=CC=CC=C45)CC3)C(=O)NC2=C1
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
  • (1)
Industry Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)