Learn More
Domperidone 1.0 mg/ml in Methanol, LoGiCal
Product Code. 30458392
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458392 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30458392 Supplier LoGiCal Supplier No. LGCAMP0107.0001
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Domperidone 1.0 mg/ml in Methanol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Domperidone
CAS 57808-66-9
Analyte or Component Names Domperidone
Shipping Condition Room Temperature
Molecular Formula C22 H24 Cl N5 O2
Certifications/Compliance ISO 17034
Synonym Domperidone
InChI Formula InChI=1 S/C22H24ClN5O2/c23-15-6-7-20-18(14-15)25-22(30)28(20)16-8-12-26(13-9-16)10-3-11-27-19-5-2-1-4-17(19)24-21(27)29/h1-2,4-7,14,16 H,3,8-13H2,(H,24,29)(H,25,30)
SMILES Clc1ccc2N(C3CCN(CCCN4C(=O)Nc5ccccc45)CC3)C(=O)Nc2c1
IUPAC Name 6-chloro-3-[1-[3-(2-oxo-3 H-benzimidazol-1-yl)propyl]piperidin-4-yl]-1 H-benzimidazol-2-one
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 425.91
Quantity 1 mL
Formula Weight 425.1619 g/mol