accessibility menu, dialog, popup

UserName

CAS RN 536-78-7

H3CN

CAS RN 536-78-7

IUPAC Name: 3-ethylpyridine
Synonyms: 3-ethylpyridine
Molecular Weight (g/mol): 107.16
Molecular Formula: C7H9N
InChi Key: MFEIKQPHQINPRI-UHFFFAOYSA-N
SMILES: CCC1=CC=CN=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results