missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethylpyridine, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
3-Ethylpyridine
Specifications
Specifications
| Chemical Name or Material | 3-Ethylpyridine |
| CAS | 536-78-7 |
| Analyte or Component Names | 3-Ethylpyridine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H9 N |
| Certifications/Compliance | ISO 9001 |
| Synonym | β-Ethylpyridine, β-Lutidine |
| InChI Formula | InChI=1S/C7H9N/c1-2-7-4-3-5-8-6-7/h3-6H,2H2,1H3 |
| SMILES | CCc1cccnc1 |
| IUPAC Name | 3-ethylpyridine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction