accessibility menu, dialog, popup

UserName

CAS RN 5292-21-7

OHO

CAS RN 5292-21-7

IUPAC Name: 2-cyclohexylacetic acid
Synonyms: cyclohexaneacetic acid
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: LJOODBDWMQKMFB-UHFFFAOYSA-N
SMILES: OC(=O)CC1CCCCC1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)