accessibility menu, dialog, popup

UserName

CAS RN 39620-04-7

NNH2Cl

CAS RN 39620-04-7

IUPAC Name: 3-chloropyridin-2-amine
Synonyms: 3-chloropyridin-2-amine
Molecular Weight (g/mol): 128.56
Molecular Formula: C5H5ClN2
InChi Key: RZJPBQGRCNJYBU-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=CC=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)