Learn More
2-Amino-3-chloropyridine, 97%, Thermo Scientific™
Product Code. 15299856 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15299856
Brand: Thermo Scientific Acros 453710010
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Chemical Identifiers
CAS 39620-04-7
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.56
InChI Key RZJPBQGRCNJYBU-UHFFFAOYSA-N
Synonym 2-amino-3-chloropyridine, 3-chloro-pyridin-2-ylamine, 2-amino-3-chloro pyridine, 3-chloro-2-pyridinamine, 3-chloro-2-pyridinylamine, 2-pyridinamine, 3-chloro, 3-chloro-2-pyridylamine, 2-amino-3-chloro-pyridine, 3-chloranylpyridin-2-amine, o-aminochloropyridine
PubChem CID 693267
IUPAC Name 3-chloropyridin-2-amine
SMILES C1=CC(=C(N=C1)N)Cl
Specifications
Melting Point 58.0°C to 60.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 128.56
Percent Purity 97%
Chemical Name or Material 2-Amino-3-chloropyridine
compliance-icons
Product Identifier
  • 2-Amino-3-chloropyridine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.