CAS RN 3794-83-0
CAS RN 3794-83-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(1-Hydroxyethylidene)bis-phosphonic Acid Tetrasodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(1-Hydroxyethylidene)bis-phosphonic Acid Tetrasodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(1-Hydroxyethylidene)bis-phosphonic Acid Tetrasodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.