missing translation for 'onlineSavingsMsg'
Learn More

(1-Hydroxyethylidene)bis-phosphonic Acid Tetrasodium Salt, TRC

Product Code. 30389492
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389492 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389492 Supplier Toronto Research Chemicals Supplier No. TRCH5383901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1-Hydroxyethylidene)bis-phosphonic Acid Tetrasodium Salt
CAS 3794-83-0
Molecular Formula C2 H4 O7 P2 . 4 Na
Quantity 1 g
Synonym (Hydroxyethylidene)diphosphonic Acid Tetrasodium Salt, 1-Hydroxyethane-1,1-diphosphonic acid Tetrasodium Salt, 1-Hydroxyethylidene-1,1-diphosphonic Acid Tetrasodium Salt, Briquest ADPA 21SH, Chelest PH 214, DQ 2016, Defloc EN 43, Dequest 2016, Dequest 2016D, Encap 81105, Etidronic Acid Tetrasodium Salt, Feliox 115A Tetrasodium Salt, PH 214, SHEDN, Sequion 10Na4, Tarpinel 4NL, Tetrasodium (1-hydroxyethylidene)bis[phosphonate], Tetrasodium (1-hydroxyethylidene)diphosphonate, Tetrasodium 1-hydroxy-1,1-ethanediphosphonate, Tetrasodium HEDP, Tetrasodium Etidronate, Tetrasodium hydroxyethylidene-1,1-diphosphonate, Turpinal 4NL
InChI Formula InChI=1S/C2H8O7P2.4Na/c1-2(3,10(4,5)6)11(7,8)9;;;;/h3H,1H3,(H2,4,5,6)(H2,7,8,9);;;;/q;4*+1/p-4
SMILES [Na+].[Na+].[Na+].[Na+].CC(O)(P(=O)([O-])[O-])P(=O)([O-])[O-]
IUPAC Name tetrasodium;1,1-diphosphonatoethanol
Molecular Weight (g/mol) 293.956
Formula Weight 293.902
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.