missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-Hydroxyethylidene)bis-phosphonic Acid Tetrasodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | (1-Hydroxyethylidene)bis-phosphonic Acid Tetrasodium Salt |
| CAS | 3794-83-0 |
| Molecular Formula | C2 H4 O7 P2 . 4 Na |
| Quantity | 500 mg |
| Synonym | (Hydroxyethylidene)diphosphonic Acid Tetrasodium Salt, 1-Hydroxyethane-1,1-diphosphonic acid Tetrasodium Salt, 1-Hydroxyethylidene-1,1-diphosphonic Acid Tetrasodium Salt, Briquest ADPA 21SH, Chelest PH 214, DQ 2016, Defloc EN 43, Dequest 2016, Dequest 2016D, Encap 81105, Etidronic Acid Tetrasodium Salt, Feliox 115A Tetrasodium Salt, PH 214, SHEDN, Sequion 10Na4, Tarpinel 4NL, Tetrasodium (1-hydroxyethylidene)bis[phosphonate], Tetrasodium (1-hydroxyethylidene)diphosphonate, Tetrasodium 1-hydroxy-1,1-ethanediphosphonate, Tetrasodium HEDP, Tetrasodium Etidronate, Tetrasodium hydroxyethylidene-1,1-diphosphonate, Turpinal 4NL |
| InChI Formula | InChI=1S/C2H8O7P2.4Na/c1-2(3,10(4,5)6)11(7,8)9;;;;/h3H,1H3,(H2,4,5,6)(H2,7,8,9);;;;/q;4*+1/p-4 |
| SMILES | [Na+].[Na+].[Na+].[Na+].CC(O)(P(=O)([O-])[O-])P(=O)([O-])[O-] |
| IUPAC Name | tetrasodium;1,1-diphosphonatoethanol |
| Molecular Weight (g/mol) | 293.956 |
| Formula Weight | 293.902 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?