Learn More
(1-Hydroxyethylidene)bis-phosphonic Acid Tetrasodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | (1-Hydroxyethylidene)bis-phosphonic Acid Tetrasodium Salt |
| CAS | 3794-83-0 |
| Molecular Formula | C2 H4 O7 P2 . 4 Na |
| Quantity | 5 g |
| Synonym | (Hydroxyethylidene)diphosphonic Acid Tetrasodium Salt, 1-Hydroxyethane-1,1-diphosphonic acid Tetrasodium Salt, 1-Hydroxyethylidene-1,1-diphosphonic Acid Tetrasodium Salt, Briquest ADPA 21SH, Chelest PH 214, DQ 2016, Defloc EN 43, Dequest 2016, Dequest 2016D, Encap 81105, Etidronic Acid Tetrasodium Salt, Feliox 115A Tetrasodium Salt, PH 214, SHEDN, Sequion 10Na4, Tarpinel 4NL, Tetrasodium (1-hydroxyethylidene)bis[phosphonate], Tetrasodium (1-hydroxyethylidene)diphosphonate, Tetrasodium 1-hydroxy-1,1-ethanediphosphonate, Tetrasodium HEDP, Tetrasodium Etidronate, Tetrasodium hydroxyethylidene-1,1-diphosphonate, Turpinal 4NL |
| InChI Formula | InChI=1S/C2H8O7P2.4Na/c1-2(3,10(4,5)6)11(7,8)9;;;;/h3H,1H3,(H2,4,5,6)(H2,7,8,9);;;;/q;4*+1/p-4 |
| SMILES | [Na+].[Na+].[Na+].[Na+].CC(O)(P(=O)([O-])[O-])P(=O)([O-])[O-] |
| IUPAC Name | tetrasodium;1,1-diphosphonatoethanol |
| Molecular Weight (g/mol) | 293.956 |
| Formula Weight | 293.902 |