accessibility menu, dialog, popup

UserName

CAS RN 375815-87-5

OH(S)(S)OHHOOOHOHNNN

CAS RN 375815-87-5

IUPAC Name: (2S,3S)-2,3-dihydroxybutanedioic acid; 5,8,14-triazatetracyclo[10.3.1.0²,¹¹.0⁴,⁹]hexadeca-2(11),3,5,7,9-pentaene
Synonyms: D-tartaric acid; varenicline
Molecular Weight (g/mol): 361.35
Molecular Formula: C17H19N3O6
InChi Key: TWYFGYXQSYOKLK-WUUYCOTASA-N
SMILES: O[C@@H]([C@H](O)C(O)=O)C(O)=O.C1C2CNCC1C1=C2C=C2N=CC=NC2=C1
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results