missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Varenicline Tartrate, Mikromol™
Description
Varenicline Tartrate
Specifications
Specifications
| Chemical Name or Material | Varenicline Tartrate |
| CAS | 375815-87-5 |
| Analyte or Component Names | Varenicline Tartrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H13 N3 . C4 H6 O6 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 6,10-Methano-6H-pyrazino[2,3-h][3]benzazepine, 7,8,9,10-tetrahydro-, (2R,3R)-2,3-dihydroxybutanedioate (1:1), CP 526555-18, Varenicline Tartrate |
| InChI Formula | InChI=1S/C13H13N3.C4H6O6/c1-2-16-13-5-11-9-3-8(6-14-7-9)10(11)4-12(13)15-1;5-1(3(7)8)2(6)4(9)10/h1-2,4-5,8-9,14H,3,6-7H2;1-2,5-6H,(H,7,8)(H,9,10)/t8-,9+;1-,2-/m.1/s1 |
| SMILES | O[C@H]([C@@H](O)C(=O)O)C(=O)O.C1NC[C@H]2C[C@@H]1c3cc4nccnc4cc23 |
| Molecular Weight (g/mol) | 361.35 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?