CAS RN 343787-29-1
CAS RN 343787-29-1
IUPAC Name:
1-{2-[5-(2-methoxyethoxy)-1H-1,3-benzodiazol-1-yl]quinolin-8-yl}piperidin-4-amine
Synonyms:
1-{2-[5-(2-methoxyethoxy)-1,3-benzodiazol-1-yl]quinolin-8-yl}piperidin-4-amine
Molecular Weight (g/mol):
417.51
Molecular Formula:
C24H27N5O2
InChi Key:
DEEOXSOLTLIWMG-UHFFFAOYSA-N
SMILES:
COCCOC1=CC=C2N(C=NC2=C1)C1=CC=C2C=CC=C(N3CCC(N)CC3)C2=N1