missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CP 673451
GREENER_CHOICE

Product Code. 18748208
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18748208

Brand: Tocris Bioscience™ 5993/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective PDGFR inhibitor

CP 673451 is a potent and selective PDGFR inhibitor (IC50 values are 1 and 10 nM for PDGFR-β and -α respectively). Selective for PDGFR kinase over a range of related kinases. Inhibits PDGFR-β phosphorylation in glioblastoma xenografts and PDGF-induced angiogenesis in mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[2-[5-(2-Methoxyethoxy)-1H-benzimidazol-1-yl]-8-quinolinyl]-4-piperidinamine
CAS 343787-29-1
Quantity 10 mg
Solubility Soluble to 100mM in water and to 100mM in DMSO
Molecular Formula C24H27N5O2
Formula Weight 417.5
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.