Learn More
Tocris Bioscience™ CP 673451, Tocris Bioscience™
Potent and selective PDGFR inhibitor
Brand: Tocris Bioscience™ 5993/10
This item is not returnable.
View return policy
Description
CP 673451 is a potent and selective PDGFR inhibitor (IC50 values are 1 and 10 nM for PDGFR-β and -α respectively). Selective for PDGFR kinase over a range of related kinases. Inhibits PDGFR-β phosphorylation in glioblastoma xenografts and PDGF-induced angiogenesis in mice.Specifications
| 1-[2-[5-(2-Methoxyethoxy)-1H-benzimidazol-1-yl]-8-quinolinyl]-4-piperidinamine | |
| 10 mg | |
| Solid | |
| Bioactive Small Molecule |
| 343787-29-1 | |
| Soluble to 100mM in water and to 100mM in DMSO | |
| White |